Assay Details
| Gene Symbol: ELK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257188.3 | NP_001244117.1 | 686 | 96 | ||
| XM_015127241.2 | XP_014982727.1 | 884 | 96 | |||
| GenBank mRNA | JU322442.1 | - | 819 | 96 | ||
| JU474614.1 | - | 787 | 96 | |||
| JV044587.1 | - | 856 | 96 | |||
| JV704433.1 | - | 788 | 96 | |||
More Information
Set Membership: |
|

ELK1
Genomic Map