Assay Details
| Gene Symbol: RAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266164.1 | NP_001253093.1 | 1518 | 62 | ||
| XM_028832875.1 | XP_028688708.1 | 1563 | 62 | |||
| GenBank mRNA | JV048360.1 | - | 1518 | 62 | ||
| JV490611.1 | - | 1511 | 62 | |||
More Information
Set Membership: |
|

RAB1B
Genomic Map