Assay Details
| Gene Symbol: UBE2V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266843.1 | NP_001253772.1 | 1427 | 96 | ||
| GenBank mRNA | JU336214.1 | - | 1427 | 96 | ||
| JV519048.1 | - | 1428 | 96 | |||
More Information
Set Membership: |
|

UBE2V2
Genomic Map