Assay Details
| Gene Symbol: MT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301287.1 | NP_001288216.1 | 320 | 103 | ||
| GenBank mRNA | DR774422.1 | - | 324 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MT2A| Gene Symbol: MT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301287.1 | NP_001288216.1 | 320 | 103 | ||
| GenBank mRNA | DR774422.1 | - | 324 | 103 | ||
Set Membership: |
|