Assay Details
| Gene Symbol: SKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193590.1 | NP_001180519.1 | 813 | 103 | ||
| XM_028849011.1 | XP_028704844.1 | 1011 | 103 | |||
| GenBank mRNA | CN803092.1 | - | 446 | 103 | ||
| JU470860.1 | - | 1513 | 103 | |||
| JU470861.1 | - | 826 | 103 | |||
| JV045117.1 | - | 836 | 103 | |||
| JV635639.1 | - | 867 | 103 | |||
More Information
Set Membership: |
|

SKP1
Genomic Map