Assay Details
| Gene Symbol: SERP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837183.1 | XP_028693016.1 | 296 | 73 | ||
| GenBank mRNA | JU334447.1 | - | 282 | 73 | ||
| JV047021.1 | - | 175 | 73 | |||
| JV544400.1 | - | 341 | 73 | |||
| JV683515.1 | - | 341 | 73 | |||
More Information
Set Membership: |
|

SERP2
Genomic Map