Assay Details
| Gene Symbol: CACTIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122463.2 | XP_014977949.2 | 696 | 57 | ||
| GenBank mRNA | JU476618.1 | - | 674 | 57 | ||
| JV600309.1 | - | 664 | 57 | |||
More Information
Set Membership: |
|

CACTIN
Genomic Map