Assay Details
| Gene Symbol: VSIG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846615.1 | XP_028702448.1 | 980 | 61 | ||
| GenBank mRNA | JV616391.1 | - | 209 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VSIG8| Gene Symbol: VSIG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846615.1 | XP_028702448.1 | 980 | 61 | ||
| GenBank mRNA | JV616391.1 | - | 209 | 61 | ||
Set Membership: |
|