Assay Details
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152965.2 | XP_015008451.1 | 472 | 109 | ||
| GenBank mRNA | JU325948.1 | - | 311 | 109 | ||
| JU471555.1 | - | 388 | 109 | |||
| JV578953.1 | - | 395 | 109 | |||
More Information
Set Membership: |
|

GATC
Genomic Map