Assay Details
| Gene Symbol: PCSK1N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127374.2 | XP_014982860.1 | 234 | 102 | ||
| GenBank mRNA | JV578674.1 | - | 75 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCSK1N| Gene Symbol: PCSK1N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127374.2 | XP_014982860.1 | 234 | 102 | ||
| GenBank mRNA | JV578674.1 | - | 75 | 102 | ||
Set Membership: |
|