Assay Details
| Gene Symbol: CNGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138336.2 | XP_014993822.2 | 3226 | 80 | ||
| GenBank mRNA | DQ147916.1 | - | 116 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNGA1| Gene Symbol: CNGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138336.2 | XP_014993822.2 | 3226 | 80 | ||
| GenBank mRNA | DQ147916.1 | - | 116 | 80 | ||
Set Membership: |
|