Assay Details
| Gene Symbol: GALNT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028843656.1 | XP_028699489.1 | 2581 | 74 | ||
| XM_028843657.1 | XP_028699490.1 | 2087 | 74 | |||
| XM_028843658.1 | XP_028699491.1 | 2512 | 74 | |||
| GenBank mRNA | JU332845.1 | - | 2113 | 74 | ||
| JU332846.1 | - | 2113 | 74 | |||
More Information
Set Membership: |
|

GALNT15
Genomic Map