Assay Details
| Gene Symbol: RRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139027.2 | XP_014994513.2 | 2178 | 69 | ||
| GenBank mRNA | JV705439.1 | - | 303 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RRH| Gene Symbol: RRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139027.2 | XP_014994513.2 | 2178 | 69 | ||
| GenBank mRNA | JV705439.1 | - | 303 | 69 | ||
Set Membership: |
|