Assay Details
| Gene Symbol: MGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143648.2 | XP_014999134.2 | 1604 | 117 | ||
| GenBank mRNA | JU326069.1 | - | 920 | 117 | ||
| JU473338.1 | - | 1584 | 117 | |||
| JV047096.1 | - | 1111 | 117 | |||
More Information
Set Membership: |
|

MGAT2
Genomic Map