Assay Details
| Gene Symbol: SBSPON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145564.2 | XP_015001050.1 | 942 | 91 | ||
| GenBank mRNA | JU326325.1 | - | 135 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SBSPON| Gene Symbol: SBSPON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145564.2 | XP_015001050.1 | 942 | 91 | ||
| GenBank mRNA | JU326325.1 | - | 135 | 91 | ||
Set Membership: |
|