Assay Details
| Gene Symbol: B3GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092028.4 | XP_001092028.4 | 816 | 114 | ||
| GenBank mRNA | JU326714.1 | - | 516 | 114 | ||
| JU480214.1 | - | 525 | 114 | |||
| JV706847.1 | - | 520 | 114 | |||
More Information
Set Membership: |
|

B3GALT6
Genomic Map