Assay Details
| Gene Symbol: ZBTB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836423.1 | XP_028692256.1 | 1326 | 76 | ||
| XM_028836425.1 | XP_028692258.1 | 1326 | 76 | |||
| GenBank mRNA | JU322411.1 | - | 853 | 76 | ||
| JU473355.1 | - | 858 | 76 | |||
More Information
Set Membership: |
|

ZBTB8A
Genomic Map