Assay Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002802361.3 | XP_002802407.2 | 847 | 58 | ||
| XM_015125365.2 | XP_014980851.1 | 943 | 58 | |||
| XM_015125366.2 | XP_014980852.1 | 875 | 58 | |||
| XM_015125367.2 | XP_014980853.1 | 790 | 58 | |||
| XM_015125368.2 | XP_014980854.1 | 818 | 58 | |||
| GenBank mRNA | JU333858.1 | - | 786 | 58 | ||
| JV679400.1 | - | 761 | 58 | |||
More Information
Set Membership: |
|

RAB26
Genomic Map