Assay Details
| Gene Symbol: GK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127125.2 | XP_014982611.1 | 939 | 65 | ||
| XM_015127128.2 | XP_014982614.1 | 857 | 65 | |||
| XM_015127129.2 | XP_014982615.1 | 938 | 65 | |||
| XM_015127130.2 | XP_014982616.1 | 939 | 65 | |||
| XM_015127133.2 | XP_014982619.1 | 915 | 65 | |||
| XM_015127135.2 | XP_014982621.1 | 522 | 65 | |||
| XM_028842431.1 | XP_028698264.1 | 576 | 65 | |||
| GenBank mRNA | JU333997.1 | - | 790 | 65 | ||
More Information
Set Membership: |
|

GK
Genomic Map