Assay Details
| Gene Symbol: TNFRSF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828737.1 | XP_028684570.1 | 798 | 100 | ||
| GenBank mRNA | JV048582.1 | - | 630 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF6B| Gene Symbol: TNFRSF6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828737.1 | XP_028684570.1 | 798 | 100 | ||
| GenBank mRNA | JV048582.1 | - | 630 | 100 | ||
Set Membership: |
|