Assay Details
| Gene Symbol: FAM171A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119864.2 | XP_014975350.2 | 2416 | 52 | ||
| GenBank mRNA | JU330572.1 | - | 122 | 52 | ||
| JV613834.1 | - | 2406 | 52 | |||
More Information
Set Membership: |
|

FAM171A2
Genomic Map