Assay Details
| Gene Symbol: LOC719262 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124650.2 | XP_014980136.1 | 402 | 98 | ||
| XM_028839445.1 | XP_028695278.1 | 5104 | 98 | |||
| GenBank mRNA | JU476923.1 | - | 358 | 98 | ||
More Information
Set Membership: |
|

LOC719262
Genomic Map