Assay Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112322.2 | XP_014967808.2 | 1370 | 93 | ||
| GenBank mRNA | JV050321.1 | - | 622 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STON1| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112322.2 | XP_014967808.2 | 1370 | 93 | ||
| GenBank mRNA | JV050321.1 | - | 622 | 93 | ||
Set Membership: |
|