Assay Details
| Gene Symbol: SURF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799943.3 | XP_002799989.2 | 802 | 76 | ||
| XM_028834643.1 | XP_028690476.1 | 557 | 76 | |||
| GenBank mRNA | JU323145.1 | - | 590 | 76 | ||
| JU473633.1 | - | 588 | 76 | |||
| JV048058.1 | - | 588 | 76 | |||
More Information
Set Membership: |
|

SURF1
Genomic Map