Assay Details
| Gene Symbol: CLEC2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151055.2 | XP_015006541.1 | 218 | 94 | ||
| GenBank mRNA | JU478344.1 | - | 202 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLEC2D| Gene Symbol: CLEC2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151055.2 | XP_015006541.1 | 218 | 94 | ||
| GenBank mRNA | JU478344.1 | - | 202 | 94 | ||