Assay Details
| Gene Symbol: ZBTB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808165.3 | XP_002808211.2 | 401 | 95 | ||
| GenBank mRNA | JV565135.1 | - | 127 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002808165.3 transcripts.
Set Membership: |
|

ZBTB7A
Genomic Map