Assay Details
| Gene Symbol: MRPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133668.2 | XP_014989154.1 | 604 | 81 | ||
| GenBank mRNA | JU323252.1 | - | 158 | 81 | ||
| JU473509.1 | - | 122 | 81 | |||
| JV044841.1 | - | 126 | 81 | |||
| JV601716.1 | - | 152 | 81 | |||
More Information
Set Membership: |
|

MRPS24
Genomic Map