Assay Details
| Gene Symbol: PCDH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138179.2 | XP_014993665.2 | 3643 | 76 | ||
| XM_015138183.2 | XP_014993669.2 | 4377 | 76 | |||
| GenBank mRNA | JU327136.1 | - | 2689 | 76 | ||
More Information
Set Membership: |
|

PCDH7
Genomic Map