Assay Details
| Gene Symbol: GRHPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117424.2 | XP_014972910.1 | 331 | 69 | ||
| GenBank mRNA | JU337157.1 | - | 257 | 69 | ||
| JU477885.1 | - | 223 | 69 | |||
| JV048732.1 | - | 262 | 69 | |||
| JV048733.1 | - | 262 | 69 | |||
| JV526083.1 | - | 58 | 69 | |||
More Information
Set Membership: |
|

GRHPR
Genomic Map