Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088975.4 | XP_001088975.2 | 480 | 92 | ||
| XM_015143647.2 | XP_014999133.1 | 510 | 92 | |||
| GenBank mRNA | JU474271.1 | - | 256 | 92 | ||
| JU474272.1 | - | 256 | 92 | |||
More Information
Set Membership: |
|

LRR1
Genomic Map