Assay Details
| Gene Symbol: EXOC3L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851896.1 | XP_028707729.1 | 2004 | 97 | ||
| XM_028851897.1 | XP_028707730.1 | 2004 | 97 | |||
| XM_028851898.1 | XP_028707731.1 | 728 | 97 | |||
| XM_028851899.1 | XP_028707732.1 | 1695 | 97 | |||
| XM_028851900.1 | XP_028707733.1 | 728 | 97 | |||
| XM_028851901.1 | XP_028707734.1 | 1650 | 97 | |||
| XM_028851902.1 | XP_028707735.1 | 683 | 97 | |||
| XM_028851903.1 | XP_028707736.1 | 683 | 97 | |||
| XM_028851904.1 | XP_028707737.1 | 748 | 97 | |||
| XM_028851905.1 | XP_028707738.1 | 2814 | 97 | |||
| XM_028851906.1 | XP_028707739.1 | 2814 | 97 | |||
| XM_028851907.1 | XP_028707740.1 | 554 | 97 | |||
| XM_028851908.1 | XP_028707741.1 | 2814 | 97 | |||
| GenBank mRNA | JU496215.1 | - | 115 | 97 | ||
More Information
Set Membership: |
|

EXOC3L4
Genomic Map