Assay Details
| Gene Symbol: APOL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150217.2 | XP_015005703.2 | 2153 | 110 | ||
| XM_015150218.2 | XP_015005704.2 | 2126 | 110 | |||
| XM_015150219.2 | XP_015005705.2 | 2342 | 110 | |||
| XM_015150220.2 | XP_015005706.2 | 2299 | 110 | |||
| XM_015150222.2 | XP_015005708.2 | 2275 | 110 | |||
| XM_015150223.2 | XP_015005709.2 | 2232 | 110 | |||
| XM_015150225.2 | XP_015005711.2 | 2204 | 110 | |||
| XM_015150226.2 | XP_015005712.2 | 2040 | 110 | |||
| XM_028827753.1 | XP_028683586.1 | 1813 | 110 | |||
| GenBank mRNA | JV072330.1 | - | 617 | 110 | ||
More Information
Set Membership: |
|

APOL4
Genomic Map