Assay Details
| Gene Symbol: LONRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144777.2 | XP_015000263.2 | 1547 | 81 | ||
| XM_028852361.1 | XP_028708194.1 | 1430 | 81 | |||
| GenBank mRNA | JV075380.1 | - | 923 | 81 | ||
More Information
Set Membership: |
|

LONRF1
Genomic Map