Assay Details
| Gene Symbol: LOC700344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV058529.1 | - | 1060 | 140 | ||
| JV503566.1 | - | 224 | 140 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC700344| Gene Symbol: LOC700344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV058529.1 | - | 1060 | 140 | ||
| JV503566.1 | - | 224 | 140 | |||
Set Membership: |
|