Assay Details
| Gene Symbol: BICDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841452.1 | XP_028697285.1 | 1506 | 78 | ||
| GenBank mRNA | JV606696.1 | - | 43 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BICDL2| Gene Symbol: BICDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841452.1 | XP_028697285.1 | 1506 | 78 | ||
| GenBank mRNA | JV606696.1 | - | 43 | 78 | ||
Set Membership: |
|