Assay Details
| Gene Symbol: C11H12orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152981.2 | XP_015008467.2 | 397 | 71 | ||
| XM_015152982.2 | XP_015008468.2 | 396 | 71 | |||
| GenBank mRNA | JV591118.1 | - | 250 | 71 | ||
| JV721373.1 | - | 260 | 71 | |||
More Information
Set Membership: |
|

C11H12orf43
Genomic Map