Assay Details
| Gene Symbol: PMPCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134576.2 | XP_014990062.2 | 787 | 81 | ||
| XM_028846146.1 | XP_028701979.1 | 786 | 81 | |||
| GenBank mRNA | DQ147982.1 | - | 283 | 81 | ||
| JV598970.1 | - | 735 | 81 | |||
| JV727400.1 | - | 738 | 81 | |||
More Information
Set Membership: |
|

PMPCB
Genomic Map