Assay Details
| Gene Symbol: PLLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126244.2 | XP_014981730.2 | 83 | |||
| GenBank mRNA | JU328122.1 | - | 122 | 83 | ||
| JV508717.1 | - | 83 | ||||
More Information
Set Membership: |
|

PLLP
Genomic Map