Assay Details
| Gene Symbol: PCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799285.3 | XP_002799331.2 | 294 | 100 | ||
| GenBank mRNA | JU334752.1 | - | 221 | 100 | ||
| JU334753.1 | - | 221 | 100 | |||
| JU475794.1 | - | 172 | 100 | |||
| JV043689.1 | - | 116 | 100 | |||
| JV047223.1 | - | 116 | 100 | |||
| JV583341.1 | - | 222 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002799285.3 transcripts.
Set Membership: |
|

PCBP1
Genomic Map