Assay Details
| Gene Symbol: RARB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132297.2 | XP_014987783.1 | 2307 | 65 | ||
| GenBank mRNA | JV553502.1 | - | 412 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RARB| Gene Symbol: RARB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132297.2 | XP_014987783.1 | 2307 | 65 | ||
| GenBank mRNA | JV553502.1 | - | 412 | 65 | ||
Set Membership: |
|