Assay Details
| Gene Symbol: MRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835840.1 | XP_028691673.1 | 531 | 87 | ||
| GenBank mRNA | JU335253.1 | - | 734 | 87 | ||
| JU476223.1 | - | 742 | 87 | |||
| JV047519.1 | - | 733 | 87 | |||
| JV620980.1 | - | 755 | 87 | |||
| JV636251.1 | - | 743 | 87 | |||
More Information
Set Membership: |
|

MRM3
Genomic Map