Assay Details
| Gene Symbol: USH1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120289.2 | XP_014975775.1 | 3859 | 98 | ||
| GenBank mRNA | JV642005.1 | - | 718 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

USH1G| Gene Symbol: USH1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120289.2 | XP_014975775.1 | 3859 | 98 | ||
| GenBank mRNA | JV642005.1 | - | 718 | 98 | ||
Set Membership: |
|