Assay Details
| Gene Symbol: PSMC3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808127.3 | XP_002808173.2 | 1631 | 74 | ||
| XM_028836513.1 | XP_028692346.1 | 1631 | 74 | |||
| GenBank mRNA | JU334203.1 | - | 185 | 74 | ||
| JV659917.1 | - | 2620 | 74 | |||
More Information
Set Membership: |
|

PSMC3IP
Genomic Map