Assay Details
| Gene Symbol: LOC720206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU328659.1 | - | 119 | 131 | ||
| JU328660.1 | - | 119 | 131 | |||
| JV707631.1 | - | 127 | 131 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC720206| Gene Symbol: LOC720206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU328659.1 | - | 119 | 131 | ||
| JU328660.1 | - | 119 | 131 | |||
| JV707631.1 | - | 127 | 131 | |||
Set Membership: |
|