Assay Details
Gene Symbol: NRARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028834544.1 | XP_028690377.1 | 643 | 64 | ||
GenBank mRNA | JU328084.1 | - | 260 | 64 | ||
JU473356.1 | - | 322 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
