Assay Details
| Gene Symbol: LOC722798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124914.2 | XP_014980400.1 | 1943 | 88 | ||
| GenBank mRNA | JV493376.1 | - | 126 | 88 | ||
| Gene Symbol: RDH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028838570.1 | XP_028694403.1 | 1959 | 88 | ||
| GenBank mRNA | JV706066.1 | - | 524 | 88 | ||
More Information
Set Membership: |
|

LOC722798
Genomic Map