Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131198.2 | XP_014986684.2 | 2795 | 134 | ||
| XM_028844017.1 | XP_028699850.1 | 2740 | 134 | |||
| XM_028844018.1 | XP_028699851.1 | 2547 | 134 | |||
| XM_028844019.1 | XP_028699852.1 | 2214 | 134 | |||
| GenBank mRNA | JV656014.1 | - | 924 | 134 | ||
More Information
Set Membership: |
|

ZKSCAN7
Genomic Map