Assay Details
| Gene Symbol: IER5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116647.2 | XP_014972133.2 | 250 | 77 | ||
| GenBank mRNA | JU324698.1 | - | 215 | 77 | ||
| JV683646.1 | - | 221 | 77 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015116647.2 transcripts.
Set Membership: |
|

IER5L
Genomic Map