Assay Details
| Gene Symbol: ANGPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097757.4 | XP_001097757.2 | 5191 | 89 | ||
| GenBank mRNA | JV712744.1 | - | 1525 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANGPT2| Gene Symbol: ANGPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097757.4 | XP_001097757.2 | 5191 | 89 | ||
| GenBank mRNA | JV712744.1 | - | 1525 | 89 | ||
Set Membership: |
|