Assay Details
Gene Symbol: TULP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137047.2 | XP_014992533.2 | 6571 | 102 | ||
XM_028847293.1 | XP_028703126.1 | 6503 | 102 | |||
GenBank mRNA | JU326511.1 | - | 1963 | 102 | ||
JV719186.1 | - | 1980 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
